Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Fluoribacter bozemanae derived from ATCC 33217
SDP

Catalog No. 23047289 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-289 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-289 Supplier Microbiologics™ Supplier No. 0212L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent and you’re ready to inoculate your culture media. LYFO DISK, packaged in a glass vial containing 6 pellets of a single strain, offers a ton of flexibility.

  • Biosafety Level: 2
  • 6 lyophilized pellets of a single microorganism strain for quality control

Specifications

ATCC® ATCC 33217
Biosafety Level 2
Content And Storage 2°C to 8°C
Format LYFO DISK
Manufacturing Origin United States
Product Type Lyfo-Disk QC Control
For Use With (Application) Microbial Identification
Packaging Type Glass Vial
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.