Learn More
ATCC Hoylesella oralis
Catalog No. 502379956
Encompass
Hoylesella oralis strain VPI D27B-24 is a whole-genome sequenced bacterial type strain that was isolated from a periodontal pocket.
Catalog No. 50-237-9956 Supplier ATCC Supplier No. 33269
Hoylesella oralis strain VPI D27B-24 is a whole-genome sequenced bacterial type strain that was isolated from a periodontal pocket.