Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Fluoride 100 ppm w/v in 50% v/v Tisab Buffer (1L)
SDP

Catalog No. NC0043706
Encompass_Preferred

Fluoride 100 ppm w/v in 50% v/v Tisab Buffer (1L)

Catalog No. NC0043706 Supplier Aqua Solutions Supplier No. AM348
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluoride 100 ppm w/v in 50% v/v Tisab Buffer (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.