Learn More
Hach Company Fluoride Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501994810 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Fluoride determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Fluoride level readings. Eliminate most interferences. Pack of 100 powder pillows.
Catalog No. 50-199-4810 Supplier Hach Company Supplier No. 258999
Ionic Strength Adjustor (ISA) for Fluoride determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Fluoride level readings. Eliminate most interferences. Pack of 100 powder pillows.