Learn More
Hach Company Fluoride Ionic Strength Adjustor (ISA), 454 g
Supplier:  Hach Company 258901
Encompass
Fluoride Total Ionic Strength Adjustor Buffer (TISAB) for use in fluoride determinations by electrode methods. Eliminates interferences from aluminum and iron. Powder form does not dilute samples. Ideal for determining very low fluoride levels. 454 g jar.
Catalog No. 50-199-4809