Learn More
ERA WATERS CORP FLUORIDE CAL STAND 125ML
SDP
Catalog No. NC0155115
Encompass
One 125 mL standard containing Fluoride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC0155115 Supplier ERA WATERS CORP Supplier No. 050
One 125 mL standard containing Fluoride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.