Learn More
Hach Company Fluoride Chemkey Reagents, 0.10-4.0 mg/L as F, Box of 25 - For Portable Parallel Analyzer
Catalog No. NC3036582 Shop All Hach Company Products
Encompass
Fluoride Chemkey reagents are integrated, single-use reagent tests for portable parallel analyzers used to determine fluoride in water. The chemistry follows Method 10312 and is USEPA approved for reporting in drinking water analysis. Ensure the analyzer software is updated to the required version before use.
  • Measurement range: 0.10-4.0 mg/L as F.
  • Pack quantity: 25 tests per box.
  • Intended instrument: portable parallel analyzers.
  • Method: Method 10312 (USEPA approved).
  • Applications: municipal water and potable water monitoring.
  • Simplifies testing by providing integrated reagents and eliminating manual reagent preparation.
Catalog No. NC3036582 Supplier Hach Company Supplier No. 9878000
Fluoride Chemkey reagents are integrated, single-use reagent tests for portable parallel analyzers used to determine fluoride in water. The chemistry follows Method 10312 and is USEPA approved for reporting in drinking water analysis. Ensure the analyzer software is updated to the required version before use.
  • Measurement range: 0.10-4.0 mg/L as F.
  • Pack quantity: 25 tests per box.
  • Intended instrument: portable parallel analyzers.
  • Method: Method 10312 (USEPA approved).
  • Applications: municipal water and potable water monitoring.
  • Simplifies testing by providing integrated reagents and eliminating manual reagent preparation.