Learn More
SPEX CERTIPREP LLC 3P FLUORIDE IC STD 100PPM 100ML
Catalog No. NC9408382
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Fluoride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9408382 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICFF100
(HPS by Zeptometrix) 100 ug/mL Fluoride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.