Learn More
Fluoride in Impinger Solution, Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111004500 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
Catalog No. Quantity
11-100-4500 20 mL
1 options
Catalog No. 11-100-4500 Supplier MilliporeSigma Supelco Supplier No. PEA190220ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20 mL
Concentration or Composition (by Analyte or Components) Fluoride 1-50 mg/dscm