Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride ion Solution, For ISE 0.1M F- (for ion-selective electrodes), MilliporeSigma™ Supelco™

Catalog No. 111015468 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5468 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5468 Supplier MilliporeSigma Supelco Supplier No. 47072100ML
Add to Cart
Add to Cart

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7681-49-4
Density 1.005 g/mL (at 25°C)
Linear Formula NaF
MDL Number MFCD00003524
For Use With (Equipment) Ion Selective Electrode
Synonym Sodium fluoride solution
Type Analytical Standard
Quantity 100 mL
Formula Weight 41.99
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 0.1 M F -
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.