Learn More
Hach Company Fluoride Ionic Strength Adjustor (ISA), 3.78 L
Supplier:  Hach Company 2829017
Encompass
Fluoride Total Ionic Strength Adjustor Buffer Solution (TISAB) for fluoride determinations by electrode methods. Eliminates interferences from aluminum and iron.
Catalog No. 50-199-6731