Learn More
Inorganic Ventures ICP FLUORIDE GRP STD 125mL
SDP
Supplier:  Inorganic Ventures CCS5125ML
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Analyte: Sb, Ge, Mo, P, Si, Ta, Ti, Zr, B, Hf, Nb, Re, S, Sn, and W 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-178