Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 10 ppm F, Reagents

Catalog No. 01335233
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-233 1 L
01-335-230 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-233 Supplier Reagents Holdings Llc Supplier No. CF1349001A
Add to Cart
Add to Cart

Fluoride Standard, 10 ppm F, Reagent, use for Water analysis (Fluoride content). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fluoride Standard, 10 ppm F
Packaging Poly Bottle
CAS 7732-18-5
Specific Gravity 1.0001
Physical Form Liquid
Molecular Formula NaF/H2O
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Fluoride Meter
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.