Learn More
Fluoride Standard, 1000μg/mL in water, Ultra Scientific
Specifications
Specifications
| CAS Max % | 99.9, 0.1 |
| Analyte or Component Names | Water,fluoride,soluble compounds as F |
| Element | Fluoride, Soluble compounds as F |
| Solvent or Matrix | Water |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
| Chemical Name or Material | Fluoride Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.