Learn More
AccuStandard, Inc FLUORIDE STD 100ML
SDP
Catalog No. NC9984688
Encompass
Anion Standard - Fluoride. 100 mL. 10 �g/mL in 2-5% Nitric Acid.
Catalog No. NC9984688 Supplier AccuStandard, Inc Supplier No. ICF1X1
Anion Standard - Fluoride. 100 mL. 10 �g/mL in 2-5% Nitric Acid.