Learn More
Hach Company Fluoride Standard Solution, 1 mg/L, 1 L
Supplier:  Hach Company 29153
Encompass
ISE Standard Solution. Fluoride, 1.00 mg/L ±0.02 mg/L as F- (NIST). 1 L bottle
Catalog No. 50-199-4236