Learn More
Starna Cells Inc FLUORIMETER CELLS UV QUARTZ
SDP
Catalog No. NC9520499
Encompass_Preferred
Fluorometer Cells Rectangular Pathlength:10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width:10mm Interior Length: 10mm Nominal Volume: 3.5mL Number polished windows: 4 Window Material: Spectrosil Quartz
Catalog No. NC9520499 Supplier Starna Cells Inc Supplier No. 3Q10
Fluorometer Cells Rectangular Pathlength:10mm Exterior Width: 12.5mm Exterior Length: 12.5mm Exterior Height: 45mm Interior Width:10mm Interior Length: 10mm Nominal Volume: 3.5mL Number polished windows: 4 Window Material: Spectrosil Quartz