Learn More
Starna Cells Inc FLUORIMETER MICROCELL 5MM
SDP
Catalog No. NC9747224
Encompass_Preferred
Square Micro Fluorometer Cell,open top,Window material: Spectrosil Quartz or equivalent, useable range: 170 to 2700 nm,Path Length:5 mm, Exterior (W x L x H): 7.5 x 7.5 x 45 mm, Interior(W x L): 5 x 5 mm ,Nominal Volume: 0.875 ml, Number Po
Catalog No. NC9747224 Supplier Starna Cells Inc Supplier No. 35.45Q5
Square Micro Fluorometer Cell,open top,Window material: Spectrosil Quartz or equivalent, useable range: 170 to 2700 nm,Path Length:5 mm, Exterior (W x L x H): 7.5 x 7.5 x 45 mm, Interior(W x L): 5 x 5 mm ,Nominal Volume: 0.875 ml, Number Po