Learn More
Sigma Aldrich Fine Chemicals Biosciences fluorimetric Hydrogen Pero
Catalog No. 502904427
Encompass
Hydrogen peroxide a reactive oxygen species produced through the metabolism of molecular oxygen serves as both an intracellular signaling messenger and a source of oxidative stress. Hydrogen peroxide is generated in cells via multiple mechanisms such as the NOX-mediated ROS production by neutrophils and macrophages (respiratory burst) or by the dismutase of superoxide anions produced as a result of electron leak during mitochondrial respiration. Abnormal hydrogen peroxide production contributes to oxidative cell damage and the progression of diseases such as asthma atherosclerosis osteoporosis and neurodegeneration.
Catalog No. 50-290-4427 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1651KT
Hydrogen peroxide a reactive oxygen species produced through the metabolism of molecular oxygen serves as both an intracellular signaling messenger and a source of oxidative stress. Hydrogen peroxide is generated in cells via multiple mechanisms such as the NOX-mediated ROS production by neutrophils and macrophages (respiratory burst) or by the dismutase of superoxide anions produced as a result of electron leak during mitochondrial respiration. Abnormal hydrogen peroxide production contributes to oxidative cell damage and the progression of diseases such as asthma atherosclerosis osteoporosis and neurodegeneration.