Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorinert™ Perfluorotributylamine (PFTBA), MilliporeSigma™ Supelco™

Catalog No. 111002174 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2174 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2174 Supplier MilliporeSigma Supelco Supplier No. 442747U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line Fluorinert
CAS 311-89-7
Vapor Pressure 1.3 mmHg (25°C)
Vapor Density 23.3 (vs. air)
Density 1.883 g/mL (at 25°C)
Physical Form Neat
Linear Formula N(CF2 CF2 CF2 CF3)3
MDL Number MFCD00000436
Boiling Point 178°C
Synonym Heptacosafluorotributylamine; Fluorinert(TM) FC-43; PFTBA; Perfluorotributylamine; Tris(nonafluorobutyl)amine
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 671.09
Grade Analytical Standard
Show More Show Less

Fluorinert is a trademark of 3M Company

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.