Learn More
Medchemexpress LLC Fluorizoline 10mg | 1362243-70-6 | 10 MG
SDP
Catalog No. 5000307879
Encompass_Preferred
Fluorizoline is a small-molecule research compound that induces apoptosis by selectively binding prohibitin proteins (PHB1 and PHB2) and upregulating pro-apoptotic factors such as NOXA and BIM. It is supplied as a high-purity solid and as a DMSO solution for use in in vitro cellular and biochemical studies.
  • Induces apoptosis via prohibitin 1 and 2 binding.
  • High purity (99.77%).
  • Available in small solid quantities and as a 10 mM DMSO solution.
  • White to off-white solid appearance for easy identification.
  • Stable when stored under recommended conditions.
Catalog No. 50-003-07879 Supplier Medchemexpress LLC Supplier No. HY11498910MG
Fluorizoline is a small-molecule research compound that induces apoptosis by selectively binding prohibitin proteins (PHB1 and PHB2) and upregulating pro-apoptotic factors such as NOXA and BIM. It is supplied as a high-purity solid and as a DMSO solution for use in in vitro cellular and biochemical studies.
  • Induces apoptosis via prohibitin 1 and 2 binding.
  • High purity (99.77%).
  • Available in small solid quantities and as a 10 mM DMSO solution.
  • White to off-white solid appearance for easy identification.
  • Stable when stored under recommended conditions.