Learn More
Medchemexpress LLC Fluorizoline | 25MG
SDP
Catalog No. 5000228486
Encompass_Preferred
Fluorizoline selectively and directly binds to prohibitin 1 (PHB1) and 2 (PHB2) and induces apoptosis Fluorizoline reduces chronic lymphocytic leukemia (CLL) cell viability through the upregulation of NOXA and BIM Fluorizoline exerts antitumor action in a p53-independent manner[1]
Catalog No. 50-002-28486 Supplier Medchemexpress LLC Supplier No. HY11498925MG
Fluorizoline selectively and directly binds to prohibitin 1 (PHB1) and 2 (PHB2) and induces apoptosis Fluorizoline reduces chronic lymphocytic leukemia (CLL) cell viability through the upregulation of NOXA and BIM Fluorizoline exerts antitumor action in a p53-independent manner[1]