Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorizoline 50mg | 1362243-70-6 | 362.20 g/mol | C15H8Cl2F3NS | 50 MG
SDP

Catalog No. 5000226840
Encompass_Preferred

Fluorizoline is a small-molecule antitumor agent that selectively binds prohibitin 1 (PHB1) and prohibitin 2 (PHB2) to induce apoptosis. It reduces chronic lymphocytic leukemia (CLL) cell viability through upregulation of NOXA and BIM and demonstrates activity independent of p53. The compound is supplied as a solid powder for preclinical research.

  • High purity (99.77%) suitable for research assays.
  • Molecular weight 362.20 g/mol and formula C15H8Cl2F3NS.
  • Soluble in DMSO (50 mg/mL); may require ultrasonic treatment.
  • Powder stable at -20°C for long-term storage.
  • Direct binder of prohibitin proteins; useful for apoptosis and mechanism-of-action studies.

Catalog No. 50-002-26840 Supplier Medchemexpress LLC Supplier No. HY11498950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorizoline is a small-molecule antitumor agent that selectively binds prohibitin 1 (PHB1) and prohibitin 2 (PHB2) to induce apoptosis. It reduces chronic lymphocytic leukemia (CLL) cell viability through upregulation of NOXA and BIM and demonstrates activity independent of p53. The compound is supplied as a solid powder for preclinical research.

  • High purity (99.77%) suitable for research assays.
  • Molecular weight 362.20 g/mol and formula C15H8Cl2F3NS.
  • Soluble in DMSO (50 mg/mL); may require ultrasonic treatment.
  • Powder stable at -20°C for long-term storage.
  • Direct binder of prohibitin proteins; useful for apoptosis and mechanism-of-action studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.