Learn More
Biocare Medical, LLC Fluoro Care Anti-Fade Mountant - 10 ml
SDP
Supplier:  Biocare Medical, LLC FP001G10
Encompass_Preferred
Fluoro Care Anti-Fade Mountant is an innovative, liquid-based, anti-fade mounting medium designed especially for the permanent preservation of fluorescent specimens. Fluoro Care is to be used with tissue sections stained with a multitude of fluorescent dyes. Fluoro Care is compatible with DyLightTM Fluors, Alexa Fluors®, fluorescein, rhodamine, Texas Red and stained specimens.
Catalog No. 50-828-50