Learn More
Cayman Chemical fluoro-DapaglIflozIn 10mg
SDP
Catalog No. 502973254
Encompass_Preferred
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively); significantly reduces phlorizin-sensitive sugar transport in COS-1 cells expressing human SGLT2
Catalog No. 50-297-3254 Supplier Cayman Chemical Supplier No. 1595110 MG
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively); significantly reduces phlorizin-sensitive sugar transport in COS-1 cells expressing human SGLT2