Learn More
Cayman Chemical fluoro-Dapagliflozin, Purity: ≥95%, CAS: 1181681-43-5, 50 mg
SDP
Catalog No. 502973052
Encompass_Preferred
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively); significantly reduces phlorizin-sensitive sugar transport in COS-1 cells expressing human SGLT2
Catalog No. 50-297-3052 Supplier Cayman Chemical Supplier No. 1595150 MG
An analog of dapagliflozin that displays similar selectivity for inhibiting SGLT2 over SGLT1 (Kis = 5.3 and 330 nM, respectively); significantly reduces phlorizin-sensitive sugar transport in COS-1 cells expressing human SGLT2