Learn More
Electron Microscopy Sciences Fluoro-Gel with Anti-Fading Agent    20 ML
SDP
Catalog No. 5033846
Encompass_Preferred
Fluoro-Gel Mounting Medium with a strong anti-fading agent 1,4-phenylenediamine is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula preserves rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa Fluoro 388, Alexa Fluoro 594 tetramethyly rhodamine, and Redox. The fluorescence is retained during prolong storage at 4 degree C in the dark.
This medium contains Phynylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PR) phycocyanin  (PC), and allophycocyanin (APC). Prepared from TRIS buffer. Ready-to-use mounting medium with dark coffee color, this is normal, and does not interfere with immuno-fluorescence.
Storage in temperature of 2-8 degree C is recommended. Protect from light. DO NOT FREEZE.
Catalog No. 50-338-46 Supplier Electron Microscopy Sciences Supplier No. 1798320
Fluoro-Gel Mounting Medium with a strong anti-fading agent 1,4-phenylenediamine is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula preserves rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa Fluoro 388, Alexa Fluoro 594 tetramethyly rhodamine, and Redox. The fluorescence is retained during prolong storage at 4 degree C in the dark.
This medium contains Phynylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PR) phycocyanin  (PC), and allophycocyanin (APC). Prepared from TRIS buffer. Ready-to-use mounting medium with dark coffee color, this is normal, and does not interfere with immuno-fluorescence.
Storage in temperature of 2-8 degree C is recommended. Protect from light. DO NOT FREEZE.