Learn More
Electron Microscopy Sciences Fluoro Gel with Dabco 1000 ML
SDP
Catalog No. 5034368
Encompass_Preferred
Fluoro Gel with antifading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, Cy dyes, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark. Use for immunofluorescence and confocal microscopy. Storage at 2-8 degree C is recommended
Catalog No. 50-343-68 Supplier Electron Microscopy Sciences Supplier No. 1798504
Fluoro Gel with antifading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, Cy dyes, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark. Use for immunofluorescence and confocal microscopy. Storage at 2-8 degree C is recommended