Learn More
Electron Microscopy Sciences Fluoro-Gel Mounting Medium with Tris Buffer 100 ML
SDP
Catalog No. 5024701
Encompass_Preferred
Fluoro-Gel is a water based mounting medium designed for the permanent mounting of fluorescent stained tissues, which may either be damaged or soluble in organic solvents, such as xylene or toluene. This unique formula prevents rapid photo-bleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa Fluoro 488, Alexa Fluoro 594, Green Fluorescent Protein (GFP), Tetramethylyrhodamine, Redox, Phycoerythrin (RP-E), Phyocyanin (PC), and Allophycocyanin (APC).
The fluorescence is retained during prolonged storage at 4 degree C in the dark. Application: Immunofluorescence, confocal microscope.
Catalog No. 50-247-01 Supplier Electron Microscopy Sciences Supplier No. 1798511
Fluoro-Gel is a water based mounting medium designed for the permanent mounting of fluorescent stained tissues, which may either be damaged or soluble in organic solvents, such as xylene or toluene. This unique formula prevents rapid photo-bleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa Fluoro 488, Alexa Fluoro 594, Green Fluorescent Protein (GFP), Tetramethylyrhodamine, Redox, Phycoerythrin (RP-E), Phyocyanin (PC), and Allophycocyanin (APC).
The fluorescence is retained during prolonged storage at 4 degree C in the dark. Application: Immunofluorescence, confocal microscope.