Learn More
Fluorochrome Inc Fluoro-Gold, 50 mg
Catalog No. NC0560981
Encompass
Hydroxystilbamidine (also called Fluoro-Gold) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading.
Catalog No. NC0560981 Supplier Fluorochrome Inc Supplier No. FLUOROGOLD50MG
Hydroxystilbamidine (also called Fluoro-Gold) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading.