Learn More
Histo Chem Incorporated Fluoro-Jade C Staining Kit, 10X concentrate, liquid, includes DAPI counterstain, for detecting degenerating neurons, suitable for flow cytometry and immunocytochemistry
Catalog No. NC0859611
Encompass
The Fluoro-Jade C Staining Kit is a highly specialized fluorescent system designed for the sensitive detection of degenerating neurons in various tissue types. It features Fluoro-Jade C dye, known for its exceptional signal-to-background ratio and superior imaging resolution. The kit also includes DAPI as a nuclear counterstain, aiding in the analysis of cell viability and architecture. This kit is compatible with a wide range of histological techniques and is produced in a ready-to-dilute format for user convenience.
  • High specificity for degenerating neurons
  • Optimal signal-to-background ratio
  • Integrated DAPI counterstain for cell nuclei labeling
  • Broad histological compatibility with multiple tissue types
  • Resistance to photobleaching for prolonged imaging
  • Ready-to-dilute format increases laboratory efficiency
Catalog No. NC0859611 Supplier Histo Chem Incorporated Supplier No. FJCSKDAPI
The Fluoro-Jade C Staining Kit is a highly specialized fluorescent system designed for the sensitive detection of degenerating neurons in various tissue types. It features Fluoro-Jade C dye, known for its exceptional signal-to-background ratio and superior imaging resolution. The kit also includes DAPI as a nuclear counterstain, aiding in the analysis of cell viability and architecture. This kit is compatible with a wide range of histological techniques and is produced in a ready-to-dilute format for user convenience.
  • High specificity for degenerating neurons
  • Optimal signal-to-background ratio
  • Integrated DAPI counterstain for cell nuclei labeling
  • Broad histological compatibility with multiple tissue types
  • Resistance to photobleaching for prolonged imaging
  • Ready-to-dilute format increases laboratory efficiency