Learn More
Immunochemistry Technology FLUORO NAD/NADH DETECTION KIT
Catalog No. NC9841030
Encompass
The role of pyridine nucleotides (NAD/NADH) metabolism in health, is of continual and increased interest. A growing amount of evidence supports the fact that NAD metabolism regulates important biological effect including life span. NAD, through poly-ADP-ribosyl polymerase (PARP), mono-ADP-ribosyltransferase (ARTs) and recently characterized sirtuin enzymes, exerts potential biological effects. These enzymes modify proteins to regulate their function via ADP-ribosylation or deacetylation and are involved in several pathways including apoptosis, DNA repair, senescence and endocrine signaling. This suggests that either the enzymes or NAD could be an important therapeutical target 1.
Catalog No. NC9841030 Supplier Immunochemistry Technology Supplier No. FLNADH1002
The role of pyridine nucleotides (NAD/NADH) metabolism in health, is of continual and increased interest. A growing amount of evidence supports the fact that NAD metabolism regulates important biological effect including life span. NAD, through poly-ADP-ribosyl polymerase (PARP), mono-ADP-ribosyltransferase (ARTs) and recently characterized sirtuin enzymes, exerts potential biological effects. These enzymes modify proteins to regulate their function via ADP-ribosylation or deacetylation and are involved in several pathways including apoptosis, DNA repair, senescence and endocrine signaling. This suggests that either the enzymes or NAD could be an important therapeutical target 1.