Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoroacetonitrile 98.0+%, TCI America™
SDP

Catalog No. F06655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06655G Supplier TCI America Supplier No. F06655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 503-20-8
Molecular Formula C2H2FN
Molecular Weight (g/mol) 59.043
MDL Number MFCD00009969
InChI Key GNFVFPBRMLIKIM-UHFFFAOYSA-N
PubChem CID 10420
IUPAC Name 2-fluoroacetonitrile
SMILES C(C#N)F

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 82°C
Quantity 5 g
UN Number 3273
Formula Weight 59.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluoroacetonitrile
RTECSNumber AM0175000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.