Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fluoroantimonic acid

Catalog No. 17510250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-510-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-510-250G Supplier Sigma Aldrich Supplier No. 17510250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

HF/SbF5 may be synthesized by mixing hydrogen fluoride and antimony fluoride.1 Fluoroantimonic acid (HF/SbF5), is a combination of SbF5 and HF. It is the known strongest liquid superacid.HF/SbF5 is an excellent medium for the preparation of alkylidene oxonium salts.

Specifications

CAS 16950-06-4
Density 2.885 g/mL (at 25°C)
Molecular Formula HF6Sb
Linear Formula HF -+ SbF5
Quantity 50 g
MDL Number MFCD00011327
Synonym Fluoroantimonic acid; Hexafluoroantimonic acid; Hexafluorostibonic acid; Monohydrogen hexafluoroantimonate
Molecular Weight (g/mol) 236.76
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.