Learn More
Bio Rad Laboratories FLUOROAPATITE CFT CERAMIC
Catalog No. 509013163
Encompass
CFT Ceramic Fluoroapatite; Bio-Rad; Type II; High binding capacity; Reproducible over an extended number of cycles; Chemically durable; 40um; 10g
Catalog No. 50-901-3163 Supplier Bio Rad Laboratories Supplier No. 1585200
CFT Ceramic Fluoroapatite; Bio-Rad; Type II; High binding capacity; Reproducible over an extended number of cycles; Chemically durable; 40um; 10g