Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
462-06-6
C6H5F
96.1
PYLWMHQQBFSUBP-UHFFFAOYSA-N
benzene, fluoro,monofluorobenzene,phenyl fluoride,fluorbenzol,fluoro-benzene,4-fluorobenzene,unii-g3tsz68k12,fluoranylbenzene,fluorobenzenes,1,2,3,4,5-pentadeuterio-6-fluorobenzene
10008
CHEBI:5115
fluorobenzene
C1=CC=C(C=C1)F
462-06-6
C6H5F
96.1
PYLWMHQQBFSUBP-UHFFFAOYSA-N
benzene, fluoro,monofluorobenzene,phenyl fluoride,fluorbenzol,fluoro-benzene,4-fluorobenzene,unii-g3tsz68k12,fluoranylbenzene,fluorobenzenes,1,2,3,4,5-pentadeuterio-6-fluorobenzene
10008
CHEBI:5115
fluorobenzene
C1=CC=C(C=C1)F
Specific Gravity 1.02
Infrared Spectrum Authentic
Flash Point −15°C
Water 0.1% Max. (K.F.)
Density 1.0200g/mL
Packaging Glass bottle
Color Colorless
Refractive Index 1.4643 to 1.4663
Quantity 100g
Boiling Point 85.0°C
Melting Point -42.0°C
Assay Percent Range 99%
Linear Formula C6H5F
Beilstein 05,IV,632
Merck Index 15,4201
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and water
Formula Weight 96.1
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Fluorobenzene
Show More Show Less