Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorobenzene, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019918 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9918 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9918 Supplier MilliporeSigma Supelco Supplier No. 514115MLF
Add to Cart
Add to Cart

Fluorobenzene is a colorless liquid, which can find applications as a reagent for resin or plastic polymers and also as an insecticide. It also plays the role of a capping agent in the synthesis of a non-fullerene electron acceptor like SF(DPPFB)4 comprising of a spirobifluorene core and four diketopyrrolopyrrole (DPP) arms that can find applications in designing organic solar cells.

TRUSTED_SUSTAINABILITY

Specifications

CAS 462-06-6
Density 1.024 g/mL (at 25°C)
Physical Form Neat
Linear Formula C6H5F
MDL Number MFCD00000280
Melting Point -42°C
Boiling Point 85°C
Quantity 5 mL
Formula Weight 96.10
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.