Learn More
Strem, An Ascensus Company CAS# 462-06-6. 25g. Fluorobenzene, 99%. MFCD00000280
SDP
Catalog No. 5090109312
Encompass
CAS# 462-06-6. 25g. Fluorobenzene, 99%. MFCD00000280. Molecular Weight: 96.10. Molecular Formula: C6H5F. Color/form: colorless liq. Strem# 09-3480. http://www.strem.com/catalog/v/09-3480/
Catalog No. 50-901-09312 Supplier Strem, An Ascensus Company Supplier No. 09348025G
CAS# 462-06-6. 25g. Fluorobenzene, 99%. MFCD00000280. Molecular Weight: 96.10. Molecular Formula: C6H5F. Color/form: colorless liq. Strem# 09-3480. http://www.strem.com/catalog/v/09-3480/