Learn More
Medchemexpress LLC HY-108525 5mg Medchemexpress, Fluorobexarotene CAS:1190848-23-7 Purity:>98%
SDP
Catalog No. 501968569
Encompass_Preferred
Medchemexpress, HY-108525 5mg Fluorobexarotene CAS:1190848-23-7 Fluorobexarotene (compound 20) is a potent retinoid-X-receptor (RXR) agonist, with a Ki value of 12 nM and an EC50 value of 43 nM for RXRα receptor. Fluorobexarotene possesses an apparent RXR binding affinity that is 75% greater than Bexarotene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8569 Supplier Medchemexpress LLC Supplier No. HY1085255MG
Medchemexpress, HY-108525 5mg Fluorobexarotene CAS:1190848-23-7 Fluorobexarotene (compound 20) is a potent retinoid-X-receptor (RXR) agonist, with a Ki value of 12 nM and an EC50 value of 43 nM for RXRα receptor. Fluorobexarotene possesses an apparent RXR binding affinity that is 75% greater than Bexarotene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.