Learn More
System Biosciences LLC FLUOROCET EXOSOME QUANTITATIONFluoroCet Exosome Quantitation Kit = 96 Reactions
SDP
Catalog No. NC1158462
Encompass_Preferred
Quantitation of exosomes and other extracellular vesicles with this 1 hour, antibody-free assay that has low sample input requirements. Enabling highly sensitive quantitation of exosomes in only 60 minutes, the FluoroCet™ Kit takes our popular EXOCET assay and increases the sensitivity over 30-fold. Fully compatible with all of SBI’s ExoQuick exosome isolation reagents as well as ultracentrifugation, FluoroCet is great for quantifying exosomes when samples are limiting.
Catalog No. NC1158462 Supplier System Biosciences LLC Supplier No. FCET96A1
Quantitation of exosomes and other extracellular vesicles with this 1 hour, antibody-free assay that has low sample input requirements. Enabling highly sensitive quantitation of exosomes in only 60 minutes, the FluoroCet™ Kit takes our popular EXOCET assay and increases the sensitivity over 30-fold. Fully compatible with all of SBI’s ExoQuick exosome isolation reagents as well as ultracentrifugation, FluoroCet is great for quantifying exosomes when samples are limiting.