Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorocholine chloride | 459424-38-5 | MFCD07995053 | 97.0% | 157.61 g/mol | C5H13ClFNO | 1 ML
SDP

Catalog No. 5000240636
Encompass_Preferred

Fluorocholine chloride is a research-grade biochemical reagent supplied as a ready-for-reconstitution DMSO solution at defined molarity for life-science experiments and biochemical assays. It provides a fluorinated choline analogue useful in tracer preparation and analytical studies.

  • 10 mM solution in DMSO
  • 1 mL volume
  • Purity 97%
  • Molecular formula C5H13ClFNO; molecular weight 157.61 g/mol
  • Intended for life-science research and biochemical assays
  • Ready for reconstitution and standard laboratory handling

Catalog No. 50-002-40636 Supplier Medchemexpress LLC Supplier No. HY10735110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorocholine chloride is a research-grade biochemical reagent supplied as a ready-for-reconstitution DMSO solution at defined molarity for life-science experiments and biochemical assays. It provides a fluorinated choline analogue useful in tracer preparation and analytical studies.

  • 10 mM solution in DMSO
  • 1 mL volume
  • Purity 97%
  • Molecular formula C5H13ClFNO; molecular weight 157.61 g/mol
  • Intended for life-science research and biochemical assays
  • Ready for reconstitution and standard laboratory handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.