Learn More
Cayman Chemical Fluoroclebopride, Purity: ≥98%, CAS: 154540-49-5, 10 mg
SDP
Catalog No. 502963934
Encompass_Preferred
A benzamide analog that is used in PET applications; binds dopamine receptors (Kis = 0.95, 5.7, 5.46, and 144 nM for D2(long), D3, and D4 receptors, respectively); selective for these receptors over D1, serotonin 5-HT2, and α2-adrenergic receptors (Kis = >10,000, 283, and 1,300 nM, respectively); a fluorine-18 moiety has been used to label this compound for use as a probe for studying D2/D3 receptor availability via PET in various monkey models
Catalog No. 50-296-3934 Supplier Cayman Chemical Supplier No. 2344710 MG
A benzamide analog that is used in PET applications; binds dopamine receptors (Kis = 0.95, 5.7, 5.46, and 144 nM for D2(long), D3, and D4 receptors, respectively); selective for these receptors over D1, serotonin 5-HT2, and α2-adrenergic receptors (Kis = >10,000, 283, and 1,300 nM, respectively); a fluorine-18 moiety has been used to label this compound for use as a probe for studying D2/D3 receptor availability via PET in various monkey models