Learn More
World Precision Instrument 100-Pack of FluoroDish Cell Culture Dish, 35mm Diameter, 10mm Well, Petri Dishes
SDP
Catalog No. NC1565370
Encompass
Cover-glass bottom for observing and growing cells for imaging
WPI’s FluoroDish™ tissue culture dishes provide exceptional imaging quality for many applications requiring the use of inverted microscopes such as high resolution image analysis, microinjection and electrophysical recording of fluorescent-tagged cells. Taking advantage of WPI’s extensive experience with low toxicity adhesives, FluoroDish™ petri dishes use a specially formulated adhesive that is optically clear, durable and with extremely low toxicity.
Catalog No. NC1565370 Supplier World Precision Instrument Supplier No. FD3510100
Cover-glass bottom for observing and growing cells for imaging
WPI’s FluoroDish™ tissue culture dishes provide exceptional imaging quality for many applications requiring the use of inverted microscopes such as high resolution image analysis, microinjection and electrophysical recording of fluorescent-tagged cells. Taking advantage of WPI’s extensive experience with low toxicity adhesives, FluoroDish™ petri dishes use a specially formulated adhesive that is optically clear, durable and with extremely low toxicity.