Learn More
World Precision Instrument 100-Pack of FluoroDish Cell Culture Dish, 35mm Diameter, 23mm Well, Poly-D-Lysine Coated, Petri Dishes
SDP
Catalog No. NC9662595
Encompass
Cover-glass bottom for observing and growing cells for imaging
Get the highest quality images and video for your research with FluoroDish Cell Culture dishes.  Their optical quality glass bottom is as thin as a coverslip, which ensures the least amount of distortions and excellent heat transfer without any of the autofluoresence issues so common with plastic petri dishes.  
Choose the style that suits your application. For live cell imaging, embryo research, and life science researchers working with small sample volumes, the 35mm Fluorodish with a 10mm well (FD3510) is ideal. Researchers working with expensive chemicals or experimental drugs choose the FD3510.
Catalog No. NC9662595 Supplier World Precision Instrument Supplier No. FD35PDL100
Cover-glass bottom for observing and growing cells for imaging
Get the highest quality images and video for your research with FluoroDish Cell Culture dishes.  Their optical quality glass bottom is as thin as a coverslip, which ensures the least amount of distortions and excellent heat transfer without any of the autofluoresence issues so common with plastic petri dishes.  
Choose the style that suits your application. For live cell imaging, embryo research, and life science researchers working with small sample volumes, the 35mm Fluorodish with a 10mm well (FD3510) is ideal. Researchers working with expensive chemicals or experimental drugs choose the FD3510.