Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 800 Capsules with Fluorodyne II Membrane - 0.1 m, gamma-irradiated (1/pkg)
Supplier:  Cytiva 12473
Encompass
Designed for Processing Volumes Up To 100 L-Fluorodyne II membrane is a hydrophilic polyvinylidene fluoride (PVDF) that offers higher flow rates with low protein and preservative binding, and broad chemical and temperature resistance-Suited for biological, pharmaceutical, and sterilizing filtration requirements-Scalable This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0896473