Learn More
Acton Technologies Inc FluoroEtch Safety Solvent-Pints
SDP
Catalog No. NC1547881
Encompass
Acton Technologies' proprietary, sodium-based etching solution, FluoroEtch Safety Solvent, is used for the treatment of fluoropolymers in preparation for bonding. It is used by many of the world's largest and most technically advanced companies, government institutions, leading universities, and research foundations as well as most major defense contractors in the United States also rely on the superior qualities of our etching solvent. FluoroEtch meets ASTM and mil-spec standards..
Catalog No. NC1547881 Supplier Acton Technologies Inc Supplier No. FSS100PINTS
Acton Technologies' proprietary, sodium-based etching solution, FluoroEtch Safety Solvent, is used for the treatment of fluoropolymers in preparation for bonding. It is used by many of the world's largest and most technically advanced companies, government institutions, leading universities, and research foundations as well as most major defense contractors in the United States also rely on the superior qualities of our etching solvent. FluoroEtch meets ASTM and mil-spec standards..