Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fluoroethylene carbonate

Catalog No. 90168610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-168-610G 10 g
90-168-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-168-610G Supplier Sigma Aldrich Supplier No. 90168610G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fluoroethylene carbonate (FEC) finds application as electrolyte in lithium ion batteries. The use of FEC as an electrolyte component has been shown to enable the formation of stable solid electrolyte interphase (SEI) and improve the performance of lithium ion batteries Product of Ube Industries Ltd.

Specifications

CAS 114435-02-8
Molecular Formula C3H3FO3
Linear Formula C3H3FO3
MDL Number MFCD06247543
Quantity 10 g
Synonym 4-Fluoro-1,3-dioxolan-2-one; FEC
Molecular Weight (g/mol) 106.05
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.