Learn More
Cayman Chemical Fluorofenidone, Purity: ≥98%, CAS: 848353-85-5, 50 mg
SDP
Catalog No. 502919202
Encompass_Preferred
An antifibrotic agent; inhibits angiotensin II-induced increases in TGF-β1 secretion and ROS production, as well as TGF-β1-induced production of collagen I, in NRK-52E cells at 8 mM; decreases disease severity and the renal levels of collagen I, collagen III, and CTGF in a rat model of obstructive nephropathy induced by unilateral ureteral obstruction at 500 mg/kg; increases survival in a mouse model of LPS-induced endotoxemia at 200 mg/kg
Catalog No. 50-291-9202 Supplier Cayman Chemical Supplier No. 3965150 MG
An antifibrotic agent; inhibits angiotensin II-induced increases in TGF-β1 secretion and ROS production, as well as TGF-β1-induced production of collagen I, in NRK-52E cells at 8 mM; decreases disease severity and the renal levels of collagen I, collagen III, and CTGF in a rat model of obstructive nephropathy induced by unilateral ureteral obstruction at 500 mg/kg; increases survival in a mouse model of LPS-induced endotoxemia at 200 mg/kg