Learn More
PARKER HANNIFIN CORP Fluoroflow Pleated Membrane Filter Cartridges
SDP
Catalog No. NC1757039
Encompass
The Fluoroflow pleated membrane filter cartridge is our standard product for aggressive wet etch and clean industrial chemical filtration applications. The filter provides good flow rates and on-stream life at an economical cost.
Catalog No. NC1757039 Supplier PARKER HANNIFIN CORP Supplier No. 33143100025EU
The Fluoroflow pleated membrane filter cartridge is our standard product for aggressive wet etch and clean industrial chemical filtration applications. The filter provides good flow rates and on-stream life at an economical cost.