Learn More
Bachem Abz-Gly-OH · HCl
SDP
Supplier:  Bachem 4015644.0025
Encompass_Preferred
Abz-Gly-OH · HCl, 256657-23-5, C9H10N2O3 · HCl, Mr 230.65, 25g. 2-Aminohippuric acid · HCl. Fluorogenic cleavage product of ACE substrate Abz-Gly-p-nitro-Phe-Pro.
Catalog No. 50-194-7188